Order Entry
Export
Orders LinkContactUsLinkComponent
Methanol ≥99.9% semiconductor grade

Methanol ≥99.9% semiconductor grade

4x1 L, 4
Catalog # 19393.M1
Flammable
Toxic
Health
Methanol ≥99.9% semiconductor grade
Catalog # 19393.M1
CAS Number:  67-56-1
Methanol ≥99.9% semiconductor grade
Methanol ≥99.9% semiconductor grade

4x1 L, 4

CAS Number:  67-56-1
Price
231,00 €
Log in for availability
1 * 4 items
Product Details & Documents

Purity: 99.9% min CAS No.: 67-56-1 Molecular Formula: CH3OH Formula Weight: 32.04 Flash Point: 11°(52°F) Physical Form: Liquid Density: 0.791 Refractive Index: 1.3290 Odour: Mild Melting Point: -98° Boiling Point: 64.7° Beilstein Reference: 1098229 Merck Reference: 14,5957 UN No.: 1230 MDL No.: MFCD00004595

Synonyms:  Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
  • CAS Number:67-56-1
  • Formula:CH₃OH
  • MDL Number:MFCD00004595
  • Molecular Weight:32.04 g/mol
  • Merck Index:14,5957
  • Boiling Pt:64.7 °C
  • Melting Pt:–98 °C
  • Density:0.791
  • Flash Pt:11 °C (52 °F)
  • Storage Temperature:Ambient
  • EINECS:200-659-6
  • UN:1230
  • ADR:3,II
  • REACH:01-2119433307-44
Specifications

Specifications

  • Pk:
    4
  • Pk Info:
    4x1 L

Specifications