2-Methylbutane ≥99%
4x500 ml, 4
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99+% CAS No.: 78-78-4 Molecular Formula: C5H12 Formula Weight: 72.14 Flash Point: -57°(-70°F) Physical Form: Liquid Density: 0.62 Refractive Index: 1.3540 Appearance: Colourless Boiling Point: 30° UN No.: 1265 MDL No.: MFCD00009338
Synonyms: iso-Pentane
- CAS Number:78-78-4
- Formula:CH₃CH₂CH(CH₃)₂
- MDL Number:MFCD00009338
- Molecular Weight:72.15 g/mol
- Boiling Pt:30 °C
- Melting Pt:–160 °C
- Density:0.62
- Flash Pt:–57 °C (–70 °F)
- Storage Temperature:Ambient
- EINECS:201-142-8
- UN:1265
- ADR:3,I
Documents
Specifications
Specifications
- Pk:4
- Pk Info:4x500 ml



