Order Entry
Export
Orders LinkContactUsLinkComponent
2-Methylbutane ≥99%

2-Methylbutane ≥99%

Glass bottle, 500 mL
Catalog # 19387.AP
Flammable
Health
Irritant
Environment
2-Methylbutane ≥99%
Catalog # 19387.AP
CAS Number:  78-78-4
2-Methylbutane ≥99%
2-Methylbutane ≥99%

Glass bottle, 500 mL

CAS Number:  78-78-4
Price
75,50 €
Log in for availability
1 * 500 mL
Product Details & Documents

Purity: 99+% CAS No.: 78-78-4 Molecular Formula: C5H12 Formula Weight: 72.14 Flash Point: -57°(-70°F) Physical Form: Liquid Density: 0.62 Refractive Index: 1.3540 Appearance: Colourless Boiling Point: 30° UN No.: 1265 MDL No.: MFCD00009338

Synonyms:  iso-Pentane
  • CAS Number:78-78-4
  • Formula:CH₃CH₂CH(CH₃)₂
  • MDL Number:MFCD00009338
  • Molecular Weight:72.15 g/mol
  • Boiling Pt:30 °C
  • Melting Pt:–160 °C
  • Density:0.62
  • Flash Pt:–57 °C (–70 °F)
  • Storage Temperature:Ambient
  • EINECS:201-142-8
  • UN:1265
  • ADR:3,I
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications