n-Pentane ≥98%
Glass bottle, 250 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 109-66-0 Molecular Formula: C5H12 Formula Weight: 72.15 Flash Point: -49°(-57°F) Physical Form: Liquid Density: 0.626 Refractive Index: 1.3580 Appearance: Colourless Odour: Nearly Odourless Melting Point: -130° Boiling Point: 36° Merck Reference: 14,7116 UN No.: 1265 MDL No.: MFCD00009498
Synonyms: Pentane
- CAS Number:109-66-0
- Formula:H₃C(CH₂)₃CH₃
- MDL Number:MFCD00009498
- Molecular Weight:72.15 g/mol
- Merck Index:14,7116
- Boiling Pt:36 °C
- Melting Pt:–130 °C
- Density:0.626
- Flash Pt:–49 °C (–57 °F)
- Storage Temperature:Ambient
- EINECS:203-692-4
- UN:1265
- ADR:3,II
- REACH:01-2119459286-30
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:250 mL



