Order Entry
Export
Orders LinkContactUsLinkComponent
Phenol red ACS

Phenol red ACS

Glass bottle for solids, 5 g
Catalog # 16294.06
Irritant
Phenol red ACS
Catalog # 16294.06
CAS Number:  143-74-8
Phenol red ACS
Phenol red ACS

Glass bottle for solids, 5 g

CAS Number:  143-74-8
Price
41,50 €
Log in for availability
1 * 5 g
Product Details & Documents

CAS No.: 143-74-8 Molecular Formula: C19H14O5S Formula Weight: 354.38 Physical Form: Crystalline Odour: Odourless Melting Point: 300° Beilstein Reference: 326470 Merck Reference: 14,7247 MDL No.: MFCD00003552

pH indicator

Synonyms:  3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:143-74-8
  • Formula:C₁₉H₁₄O₅S
  • MDL Number:MFCD00003552
  • Molecular Weight:354.38 g/mol
  • Merck Index:14,7247
  • Melting Pt:>300 °C
  • Storage Temperature:Ambient
  • EINECS:205-609-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications