Thymol blue ACS
Glass bottle for solids, 50 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 76-61-9 Molecular Formula: C27H30O5S Formula Weight: 466.60 Physical Form: Crystalline Melting Point: 223° dec. Solubility: Insoluble in water. Soluble in alcohol, dilute alkali solutions MDL No.: MFCD00005869
pH indicator: 1.2 (red) to 2.8 (yellow) to 9.6 (blue)
Synonyms: Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:76-61-9
- Formula:C₂₇H₃₀O₅S
- MDL Number:MFCD00005869
- Molecular Weight:466.6 g/mol
- Merck Index:14,9400
- Melting Pt:223 °C (dec.)
- Storage Temperature:Ambient
- EINECS:200-973-3
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



