Order Entry
Export
Orders LinkContactUsLinkComponent
Thymolphthalein ACS

Thymolphthalein ACS

Glass bottle for solids, 25 g
Catalog # 16245.14
Thymolphthalein ACS
Catalog # 16245.14
CAS Number:  125-20-2
Thymolphthalein ACS
Thymolphthalein ACS

Glass bottle for solids, 25 g

CAS Number:  125-20-2
Price
112,00 €
Log in for availability
1 * 25 g
Product Details & Documents

CAS No.: 125-20-2 Molecular Formula: C28H30O4 Formula Weight: 430.55 Physical Form: Crystalline Melting Point: 251-253° Solubility: Insoluble in water. Soluble in alcohol, acetone, dilute alkalies (blue Colour), H2SO4 (red Colour) Beilstein Reference: 359413 Merck Reference: 14,9401 MDL No.: MFCD00005909

pH indicator; Colourless below pH 9.3, blue above pH 10.5

Synonyms:  3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one
  • CAS Number:125-20-2
  • Formula:C₂₈H₃₀O₄
  • MDL Number:MFCD00005909
  • Molecular Weight:430.54 g/mol
  • Merck Index:14,9401
  • Melting Pt:251…253 °C
  • Storage Temperature:Ambient
  • EINECS:204-729-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications