Thymolphthalein ACS
Plastic bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 125-20-2 Molecular Formula: C28H30O4 Formula Weight: 430.55 Physical Form: Crystalline Melting Point: 251-253° Solubility: Insoluble in water. Soluble in alcohol, acetone, dilute alkalies (blue Colour), H2SO4 (red Colour) Beilstein Reference: 359413 Merck Reference: 14,9401 MDL No.: MFCD00005909
pH indicator; Colourless below pH 9.3, blue above pH 10.5
Synonyms: 3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one
- CAS Number:125-20-2
- Formula:C₂₈H₃₀O₄
- MDL Number:MFCD00005909
- Molecular Weight:430.54 g/mol
- Merck Index:14,9401
- Melting Pt:251…253 °C
- Storage Temperature:Ambient
- EINECS:204-729-7
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:5 g



