Sodium azide ≥99%
Glass bottle for solids, 10 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 26628-22-8 Molecular Formula: N3Na Formula Weight: 65.01 Physical Form: Powder or Granular Density: 1.846 Appearance: White Odour: Odourless pH: 10 Melting Point: ca 275° dec Solubility: Soluble in water and liquid ammonia. Slightly soluble in alcohol Merck Reference: 14,8581 UN No.: 1687 MDL No.: MFCD00003536
In airbag inflation, as a preservative in diagnostic medicinals
Synonyms: Sodium triazide
- CAS Number:26628-22-8
- Formula:NaN₃
- MDL Number:MFCD00003536
- Molecular Weight:65.01 g/mol
- Merck Index:14,8581
- Melting Pt:∼ 275 °C (dec.)
- Density:1.846
- Storage Temperature:Ambient
- EINECS:247-852-1
- UN:1687
- ADR:6.1,II
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



