Order Entry
Export
Orders LinkContactUsLinkComponent
 

Lead(II) nitrate ≥99.0% ACS

Plastic bottle for solids, 500 g
Catalog # 14243.36
Oxidizing
Health
Irritant
Lead(II) nitrate ≥99.0% ACS
Catalog # 14243.36
CAS Number:  10099-74-8
Lead(II) nitrate ≥99.0% ACS
Lead(II) nitrate ≥99.0% ACS

Plastic bottle for solids, 500 g

CAS Number:  10099-74-8
Price
63,30 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99.0% min CAS No.: 10099-74-8 Molecular Formula: Pb(NO3)2 Formula Weight: 331.20 Physical Form: Crystalline Density: 4.530 Appearance: White Odour: Odourless Melting Point: 470° dec. Solubility: Freely soluble in water. Soluble in absolute alcohol, methanol. Insoluble in concentrated HNO3 Merck Reference: 14,5414 UN No.: 1469 MDL No.: MFCD00011153

As an oxidizer in dye industry, in manufacturing of mother-of-pearl, as mordant in printing textile, as sensitizer in photography

Synonyms:  Lead dinitrate
  • CAS Number:10099-74-8
  • Formula:Pb(NO₃)₂
  • MDL Number:MFCD00011153
  • Molecular Weight:331.21 g/mol
  • Merck Index:14,5414
  • Melting Pt:470 °C (dec.)
  • Density:4.530
  • Storage Temperature:Ambient
  • EINECS:233-245-9
  • UN:1469
  • ADR:5.1,II
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g

Specifications