Order Entry
Export
Orders LinkContactUsLinkComponent
Lead(II) oxide ≥99.99% (metals basis)

Lead(II) oxide ≥99.99% (metals basis)

Plastic bottle for solids, 1 kg
Catalog # 14240.A1
Health
Irritant
Environment
Lead(II) oxide ≥99.99% (metals basis)
Catalog # 14240.A1
CAS Number:  1317-36-8
Lead(II) oxide ≥99.99% (metals basis)
Lead(II) oxide ≥99.99% (metals basis)

Plastic bottle for solids, 1 kg

CAS Number:  1317-36-8
Price
693,00 €
Log in for availability
1 * 1 kg
Product Details & Documents

Purity: 99.99% CAS No.: 1317-36-8 Molecular Formula: OPb Formula Weight: 223.19 Physical Form: Powder Density: 9.53 Appearance: Yellow Odour: Odourless Melting Point: 886° Merck Reference: 14,5413 UN No.: 3077 MDL No.: MFCD00011164

Synonyms:  Lead monoxide
  • CAS Number:1317-36-8
  • Formula:PbO
  • MDL Number:MFCD00011164
  • Molecular Weight:223.2 g/mol
  • Merck Index:14,5413
  • Boiling Pt:1470 °C (1013 hPa)
  • Melting Pt:886 °C
  • Density:9.53
  • Storage Temperature:Ambient
  • EINECS:215-267-0
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg

Specifications