Order Entry
Export
Orders LinkContactUsLinkComponent
 

Zinc selenite

Glass bottle for solids, 100 g
Catalog # 12586.22
Toxic
Health
Environment
Zinc selenite
Catalog # 12586.22
CAS Number:  13597-46-1
Zinc selenite
Zinc selenite

Glass bottle for solids, 100 g

CAS Number:  13597-46-1
Price
31,50 €
Log in for availability
1 * 100 g
Product Details & Documents

CAS No.: 13597-46-1 Molecular Formula: ZnSeO3 Formula Weight: 192.33 Physical Form: Powder UN No.: 2630 MDL No.: MFCD00011297

  • CAS Number:13597-46-1
  • Formula:O₃SeZn
  • MDL Number:MFCD00011297
  • Molecular Weight:192.33 g/mol
  • Storage Temperature:Ambient
  • EINECS:237-048-9
  • UN:2630
  • ADR:6.1,I
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications