Order Entry
Export
Orders LinkContactUsLinkComponent
 

Lead(IV) oxide ≥97.0% ACS

Glass bottle for solids, 100 g
Catalog # 12483.22
Health
Oxidizing
Irritant
Lead(IV) oxide ≥97.0% ACS
Catalog # 12483.22
CAS Number:  1309-60-0
Lead(IV) oxide ≥97.0% ACS
Lead(IV) oxide ≥97.0% ACS

Glass bottle for solids, 100 g

CAS Number:  1309-60-0
Price
85,50 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 97.0% min CAS No.: 1309-60-0 Molecular Formula: PbO2 Formula Weight: 239.19 Physical Form: Powder Density: 9.38 Melting Point: 290° dec. Solubility: Insoluble in water. Soluble in HCl (with evolution of chlorine), oxalic acid, HNO3 in presence of H2O2, alkali iodide solutions, hot alkali caustic solutions Merck Reference: 14,5407 UN No.: 1872 MDL No.: MFCD00011165

In battery electrodes, manufacturing of pigments and dyes, as analytical lab reagent

Synonyms:  Lead dioxide
  • CAS Number:1309-60-0
  • Formula:O₂Pb
  • MDL Number:MFCD00011165
  • Molecular Weight:239.2 g/mol
  • Merck Index:14,5407
  • Melting Pt:290 °C (dec.)
  • Density:9.38
  • Storage Temperature:Ambient
  • EINECS:215-174-5
  • UN:1872
  • ADR:5.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications