Order Entry
Export
Orders LinkContactUsLinkComponent
Triethylamine ≥99%

Triethylamine ≥99%

Glass bottle for solids, 250 g
Catalog # 12391.30
Flammable
Corrosive
Irritant
Triethylamine ≥99%
Catalog # 12391.30
CAS Number:  121-44-8
Triethylamine ≥99%
Triethylamine ≥99%

Glass bottle for solids, 250 g

CAS Number:  121-44-8
Price
64,00 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 99+% CAS No.: 121-44-8 Molecular Formula: C6H15N Formula Weight: 101.19 Flash Point: -11°(12°F) Physical Form: Liquid Density: 0.726 Refractive Index: 1.4005 Appearance: Colourless Odour: Ammonia-like Melting Point: -115° Boiling Point: 89-90° Beilstein Reference: 1843166 Merck Reference: 14,9666 UN No.: 1296 MDL No.: MFCD00009051

Synonyms:  (Diethylamino)ethane, TEA
  • CAS Number:121-44-8
  • Formula:(C₂H₅)₃N
  • MDL Number:MFCD00009051
  • Molecular Weight:101.19 g/mol
  • Merck Index:14,9666
  • Boiling Pt:89…90 °C
  • Melting Pt:–115 °C
  • Density:0.726
  • Flash Pt:–11 °C (–12 °F)
  • Storage Temperature:Ambient
  • EINECS:204-469-4
  • UN:1296
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications