Order Entry
Export
Orders LinkContactUsLinkComponent
 

Zinc iodide ≥98%

Plastic bottle for solids, 250 g
Catalog # 11661.30
Corrosive
Zinc iodide ≥98%
Catalog # 11661.30
CAS Number:  10139-47-6
Zinc iodide ≥98%
Zinc iodide ≥98%

Plastic bottle for solids, 250 g

CAS Number:  10139-47-6
Price
413,00 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Zinc (II) iodide
  • CAS Number:10139-47-6
  • Formula:I₂Zn
  • MDL Number:MFCD00011299
  • Molecular Weight:319.2 g/mol
  • Merck Index:14,10140
  • Boiling Pt:625 °C (dec.)
  • Melting Pt:446 °C
  • Density:4.74
  • Storage Temperature:Ambient
  • EINECS:233-396-0
  • UN:3260
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g

Specifications