Order Entry
Export
Orders LinkContactUsLinkComponent
VWR®, Conical Sample Containers, with Snap-On Lids

VWR®, Conical Sample Containers, with Snap-On Lids

400 ml, 100 mm, Assembled snap-on lid, sterile, 85/72 mm
Catalog # 216-2652
Supplier:   VWR Collection
VWR®, Conical Sample Containers, with Snap-On Lids
Catalog # 216-2652
Supplier:  VWR Collection
VWR®, Conical Sample Containers, with Snap-On Lids
VWR®, Conical Sample Containers, with Snap-On Lids

400 ml, 100 mm, Assembled snap-on lid, sterile, 85/72 mm

Price
266,00 €
Log in for availability
1 * 185 items
Product Details & Documents

PP, translucent.

  • Available in sterile or non sterile versions
  • Raised graduations
  • Leakproof snap-on lid

Conical containers for sample collection, storage and transport.

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904.

Specifications
  • Type:
    Assembled snap-on lid, sterile
  • Capacity:
    400 ml
  • Height:
    100 mm
  • Seq:
    04
  • Ø top/Ø bottom:
    85/72 mm