Order Entry
Export
Orders LinkContactUsLinkComponent
Vented caps for Falcon® flasks

Vented caps for Falcon® flasks

Vented caps for 75 cm² flasks
Catalog # 734-1067
   Low Manufacturing Impact
Supplier:   Corning
Vented caps for Falcon® flasks
Catalog # 734-1067
Supplier:  Corning
   Low Manufacturing Impact
Vented caps for Falcon® flasks
Vented caps for Falcon® flasks

Vented caps for 75 cm² flasks

Price
153,98 €
Log in for availability
1 * 100 items
Product Details & Documents

PE with integral hydophobic 0,2 µm microporous membrane filter vent.

Vented caps minimise contamination associated with standard open incubation. Gases required for cell growth and metabolism pass freely through the vent, while microorganisms do not.

Specifications
  • Description:
    Vented caps for 75 cm² flasks
  • Cat. No.:
    734-1067
  • Environmental certification:
    ISO 14001 - Bedford
  • Environmentally Preferable:
    Low Manufacturing Impact