Pycnometers, Hubbard
Pycnometer, conical, unadjustable, 50 ml, 24/15
Catalog # NEUB0303-50-50
Supplier: NEUBERT VOLUME GLASSWAERE
Jump to:
Product Details & Documents
Borosilicate glass 3,3.
- With glass stopper
- For viscous liquids, pastes such as petroleum jelly, creams and bituminous masses
According to DIN 12806 and DIN ISO 3507.
Specifications
- NS:24/15
- Description:Pycnometer, conical, unadjustable, 50 ml
- Seq:03