Orders LinkContactUsLinkComponent
Pycnometers, Hubbard
Pycnometers, Hubbard
Pycnometer, conical, unadjustable, 25 ml, 24/15
Catalog # NEUB0303-50-25
Pycnometers, Hubbard
Catalog # NEUB0303-50-25
Pycnometers, Hubbard
Pycnometer, conical, unadjustable, 25 ml, 24/15
Price
96,30 €
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass 3,3.
  • With glass stopper
  • For viscous liquids, pastes such as petroleum jelly, creams and bituminous masses
According to DIN 12806 and DIN ISO 3507.
Specifications
  • NS:
    24/15
  • Description:
    Pycnometer, conical, unadjustable, 25 ml
  • Seq:
    02