Order Entry
Export
Orders LinkContactUsLinkComponent
 

VWR®, Transfer Pipettes, High Performance

4,0 ml, 28, 3,5 ml, 2,5 mm, 150 mm, - ml, +, 5 per bag
Catalog # 612-1689
Supplier:   VWR Collection
VWR®, Transfer Pipettes, High Performance
Catalog # 612-1689
Supplier:  VWR Collection
VWR®, Transfer Pipettes, High Performance
VWR®, Transfer Pipettes, High Performance

4,0 ml, 28, 3,5 ml, 2,5 mm, 150 mm, - ml, +, 5 per bag

Price
80,00 €
Log in for availability
1 * 500 items
Product Details & Documents

LDPE, disposable. These unbreakable 'all in one' pipettes eliminate the potential hazard of broken glass and exposure to infectious materials. The low affinity surface reduces the loss of cells and valuable proteins due to binding. They work well whenever there is a need for quick, safe transfer of fluids.

  • Non toxic and inert to biological fluids and most acids
  • Can be heat-sealed and frozen at very low temperatures such as in liquid nitrogen
  • Available sterile or non sterile
  • Sterility assurance level is SAL 10⁻⁶

 : Sterile pipettes are packed in medical grade paper and non toxic film.

Specifications
  • Packed:
    5 per bag
  • Sterile:
    +
  • Capacity:
    4,0 ml
  • Length:
    150 mm
  • Seq:
    23
  • Drops per ml:
    28
  • Graduation:
    - ml
  • Stem Ø:
    2,5 mm
  • Bulb draw:
    3,5 ml