Vortex mixers, Vortex-Genie®
Product Details & Documents
Rugged and reliable vortex mixers with solid metal casing and rubber feet that prevent them 'walking'. Vortex-Genie® 1 features touch-on operation and fixed speed, it is ideal for aggressive vortexing of difficult samples.
- Wide range of attachments available
- Vortex-Genie® 2T timer function; touch-on 1 to 60 seconds, hands-free 1 to 60 minutes or continuous
- Can be used in cold room or incubator
Vortex-Genie® 2 has variable speed for precise mixing from gentle to vigorous, it offers hands-free or touch-on control. Mixing action prevents spilling, even if tubes are uncapped.
Vortex-Genie® 2T has the same features as Vortex-Genie® 2, and in addition includes an integral timer function, it can also be used on continuous untimed mode.
: Vortex-Genie® 1 is supplied with quick change standard pop-off cup for single test tubes. Optional 76 mm platform suitable for beakers, flasks and multiple tests tubes is available. Vortex-Genie® 2 and 2T are supplied with standard pop-off cup for single test tubes and 76 mm platform suitable for beakers, flasks and multiple tests tubes.
: Depending on speed
Multiple sample starter set consists of a 152 mm platform, a foam insert for 60 microtubes and a foam insert for 1 standard microplate
Large sample set is ideal for mixing beakers, flasks and large tubes. The set consists of a recessed platform, 2 elastic bands, a blank foam insert, a foam insert for 4×29 to 37 mm tubes, a foam insert for 8×14 to 29 mm tubes and a foam insert for 12×9 to 16 mm tubes
Small ampoule/tube attachment enables vigorous end-to-end mixing for up to 4×15 to 17 mm Ø tubes or ampoules
Large ampoule/tube attachment enables vigorous end-to-end mixing for up to 4×10 to 17 mm Ø tubes or ampoules
Specifications
- Description:Vortex-Genie® 2 vortex mixer, EU-plug
- Shaking motion:Orbital
- Cat. No.:444-5900
- Max. capacity:500 ml*
- Orbit:4 mm
- Speed range:600 - 2700 min⁻¹
- Model:Vortex-Genie® 2
- W×D×H:122×165×165 mm
- Weight:4 kg
Accessories
Product Information
- Catalog #ForAvailabilityPrice
- Platform, 152 mm, without insert (inserts listed below)Platform, 152 mm, without insert (inserts listed below)444-5905Vortex-Genies® except Vortex-Genie® 1Log in for availability1 * 1 items - 90,40 €
Product Information
- Catalog #ForAvailabilityPrice
- Recessed platform (foam inserts listed below)Recessed platform (foam inserts listed below)444-5916Vortex-Genies® except Vortex-Genie® 1Log in for availability1 * 1 items - 244,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Elastic bands to help secure vessels onto recessed platformElastic bands to help secure vessels onto recessed platform444-5915Vortex-Genies® except Vortex-Genie® 1Log in for availability1 * 2 items - 15,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Rubber cover for Ø 75 mm platformRubber cover for Ø 75 mm platform444-5903Vortex-Genies®Log in for availability1 * 1 items - 62,90 €
Product Information
- Catalog #ForAvailabilityPrice
- Two-tier insert for 96 well plate (fits in 152 mm platform)Two-tier insert for 96 well plate (fits in 152 mm platform)444-0061Vortex-Genies® except Vortex-Genie® 1Log in for availability1 * 1 items - 126,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Large ampoule/tube attachment****Large ampoule/tube attachment****444-9162Vortex-Genies® except Vortex-Genie® 1Log in for availability1 * 1 items - 357,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Platform with rubber cover, 3 inchPlatform with rubber cover, 3 inch444-0443Vortex-Genie® 2, Vortex-Genie® pulseLog in for availability1 * 1 items - 101,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Vertical microtube holder, 15 ml and 50 ml (for 6×50 ml, 6×15 ml tubes and 12 microtubes)Vertical microtube holder, 15 ml and 50 ml (for 6×50 ml, 6×15 ml tubes and 12 microtubes)444-1052Vortex-Genie® 2, Vortex-Genie® pulseLog in for availability1 * 1 items - 352,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Assorted tube inserts: 1 each of the following: blank, 9 − 16 mm, 14 − 29 mm and 29 − 37 mm (fit in recessed platform)Assorted tube inserts: 1 each of the following: blank, 9 − 16 mm, 14 − 29 mm and 29 − 37 mm (fit in recessed platform)444-5917Vortex-Genies® except Vortex-Genie® 1Log in for availability1 * 4 items - 103,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Large sample set**Large sample set**444-0060Vortex-Genies® except Vortex-Genie® 1Log in for availability1 * 1 items - 357,00 €
Product Information
- Catalog #ForAvailabilityPrice
- TurboMix attachment for 12×1,5/2,0 ml snap-top or screw-cap microtubesTurboMix attachment for 12×1,5/2,0 ml snap-top or screw-cap microtubes444-0724Vortex Genie® 2, Vortex PulseLog in for availability1 * 1 items - 494,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Microplate adapter for applied biosystemsMicroplate adapter for applied biosystemsSCIISI-0513Vortex-Genie® 2, Vortex-Genie® pulseLog in for availability1 * 1 items - 244,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Horizontal tube holder, 50 ml (for 6×50 ml tubes)Horizontal tube holder, 50 ml (for 6×50 ml tubes)444-1043Vortex-Genie® 2, Vortex-Genie® pulseLog in for availability1 * 1 items - 255,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Small ampoule/tube attachment***Small ampoule/tube attachment***444-9161Vortex-Genies® except Vortex-Genie® 1Log in for availability1 * 1 items - 244,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Horizontal tube holder, 15 ml (for 12×15 ml tubes)Horizontal tube holder, 15 ml (for 12×15 ml tubes)444-1044Vortex-Genie® 2, Vortex-Genie® pulseLog in for availability1 * 1 items - 289,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Horizontal microtube holder (for 24 microtubes)Horizontal microtube holder (for 24 microtubes)444-1045Vortex-Genie® 2, Vortex-Genie® pulseLog in for availability1 * 1 items - 321,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Vertical high speed tube holder, 50 ml (for 3×50 ml tubes)Vertical high speed tube holder, 50 ml (for 3×50 ml tubes)444-1046Vortex-Genie® 2, Vortex-Genie® pulseLog in for availability1 * 1 items - 236,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Bottom closure with screw kitBottom closure with screw kitSCII0K-0236-408Vortex-Genie® 2, Vortex-Genie® pulseLog in for availability1 * 1 items - 89,90 €
Product Information
- Catalog #ForAvailabilityPrice
- Vertical tube holder, 15 ml (for 12×15 ml tubes)Vertical tube holder, 15 ml (for 12×15 ml tubes)444-1050Vortex-Genie® 2, Vortex-Genie® pulseLog in for availability1 * 1 items - 289,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Vertical high speed tube holder, 15 ml (for 6×15 ml tubes)Vertical high speed tube holder, 15 ml (for 6×15 ml tubes)444-1047Vortex-Genie® 2, Vortex-Genie® pulseLog in for availability1 * 1 items - 267,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Vertical high speed microtube holder (for 9 microtubes)Vertical high speed microtube holder (for 9 microtubes)444-1048Vortex-Genie® 2, Vortex-Genie® pulseLog in for availability1 * 1 items - 299,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Vertical microtube holder (for 24 microtubes)Vertical microtube holder (for 24 microtubes)444-1051Vortex-Genie® 2, Vortex-Genie® pulseLog in for availability1 * 1 items - 321,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Vertical tube holder, 50 ml (for 6×50 ml tubes)Vertical tube holder, 50 ml (for 6×50 ml tubes)444-1049Vortex-Genie® 2, Vortex-Genie® pulseLog in for availability1 * 1 items - 255,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Holder for 12×2,0 ml screw-cap microtubes (fits on TurboMix attachment)Holder for 12×2,0 ml screw-cap microtubes (fits on TurboMix attachment)444-0775Vortex-Genies® except Vortex-Genie® 1Log in for availability1 * 1 items - 145,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Foam adapter for tubes 9 − 16 mm Ø (fits in recessed platform)Foam adapter for tubes 9 − 16 mm Ø (fits in recessed platform)444-5914Vortex-Genies® except Vortex-Genie® 1Log in for availability1 * 2 items - 50,30 €
Product Information
- Catalog #ForAvailabilityPrice
- Foam insert, blank (without holes) (fits in recessed platform)Foam insert, blank (without holes) (fits in recessed platform)444-5913Vortex-Genies® except Vortex-Genie® 1Log in for availability1 * 2 items - 50,30 €
Product Information
- Catalog #ForAvailabilityPrice
- Foam adapter for tubes 14 − 29 mm Ø (fits in recessed platform)Foam adapter for tubes 14 − 29 mm Ø (fits in recessed platform)444-5912Vortex-Genies® except Vortex-Genie® 1Log in for availability1 * 2 items - 50,30 €
Product Information
- Catalog #ForAvailabilityPrice
- Foam adapter for tubes 29 − 37 mm Ø (fits in recessed platform)Foam adapter for tubes 29 − 37 mm Ø (fits in recessed platform)444-5911Vortex-Genies® except Vortex-Genie® 1Log in for availability1 * 2 items - 50,30 €
Product Information
- Catalog #ForAvailabilityPrice
- Adapter for 60 microtubes (fits in 152 mm platform)Adapter for 60 microtubes (fits in 152 mm platform)444-5921Vortex-Genies® except Vortex-Genie® 1Log in for availability1 * 2 items - 109,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Support for microtitre plates (fits in 152 mm platform)Support for microtitre plates (fits in 152 mm platform)444-5919Vortex-Genies® except Vortex-Genie® 1Log in for availability1 * 2 items - 111,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Multiple sample starter set*Multiple sample starter set*444-5904Vortex-Genies® except Vortex-Genie® 1Log in for availability1 * 1 items - 201,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Platform, Ø 75 mm without rubber coverPlatform, Ø 75 mm without rubber cover444-5902Vortex-Genies®Log in for availability1 * 1 items - 64,10 €
Product Information
- Catalog #ForAvailabilityPrice
- Replacement elastic bandsReplacement elastic bandsSCII0K-0513-900Vortex-Genie® 2, Vortex-Genie® pulseLog in for availability1 * 1 KIT - 15,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Pop-off cup
Product Information
- Catalog #ForAvailabilityPrice
- Deepwell microplate foam insert (2). Require 6-inch platformDeepwell microplate foam insert (2). Require 6-inch platformSCII504-0232-00Vortex-Genie® 2, Vortex-Genie® pulseLog in for availability1 * 2 items - 136,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Horizontal plastic clip microtube holder (for 24 microtubes)Horizontal plastic clip microtube holder (for 24 microtubes)SCIISI-HP524Vortex-Genie® 2, Vortex-Genie® pulseLog in for availability1 * 1 items - 313,00 €