Volumetric flasks, class A, blue markings, BLAUBRAND®
1000** ml, 29/32, 0,60 ± ml
Catalog # 612-7123
Supplier: Brand
Jump to:
Product Details & Documents
Borosilicate glass 3.3, with PP stopper.
- Calibrated to contain (TC, In)
- Marks and inscriptions in high contrast blue enamel
- DE-M marking
- Individual certificate included
* In addition to DIN.
**Wide neck.
Certifications: DIN EN ISO 1042
Documents
Brand Volumetric Measurement in the Laboratory
Product Literature
Specifications
- NS:29/32
- Capacity:1000** ml
- Seq:16
- Tolerance:0,60 ± ml