Order Entry
Export
Orders LinkContactUsLinkComponent
 

Instrument trays

Lids for instrument trays, 160 mm, 290 mm
Catalog # VITL79890
Supplier:   VITLAB
Instrument trays
Catalog # VITL79890
Supplier:  VITLAB
Instrument trays
Instrument trays

Lids for instrument trays, 160 mm, 290 mm

Price
16,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Made of melamine, white.

  • Tall shape
  • Rounded corners
  • Smooth surfaces, easy to clean

Practical tray for instruments, tools, and sensitive utensils. Stable and self-supporting.

 : Fitting lid made of PS, please order separately.

Specifications
  • Width:
    160 mm
  • Description:
    Lids for instrument trays
  • Length:
    290 mm
  • Seq:
    04