Orders LinkContactUsLinkComponent
Instrument trays
Instrument trays
350×250×40 mm
Catalog # 216-0287
Supplier:   VITLAB
Instrument trays
Catalog # 216-0287
Supplier:  VITLAB
Instrument trays
Instrument trays
350×250×40 mm
Price
32,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Made of melamine, white.
  • Tall shape
  • Rounded corners
  • Smooth surfaces, easy to clean
Practical tray for instruments, tools, and sensitive utensils. Stable and self-supporting.
Ordering information: Fitting lid made of PS, please order separately.
Specifications
  • Seq:
    01
  • W×D×H:
    350×250×40 mm