Tweezers
180 mm
232-1951
: VITLAB
POM, yellow. Universal tweezers for a variety of applications.
- Blunt, elastic, very good resilience
- With grooves on the outside for optimum handling and grip
- Good chemical resistance
- Length:180 mm
POM, yellow. Universal tweezers for a variety of applications.