Orders LinkContactUsLinkComponent
Forceps for cover glasses
Forceps for cover glasses
Straight, standard quality, 115 mm
Catalog # 232-2181
Supplier:   RSG
Forceps for cover glasses
Catalog # 232-2181
Supplier:  RSG
Forceps for cover glasses
Forceps for cover glasses
Straight, standard quality, 115 mm
Price
10,90 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel.
  • For easy, secure handling of cover glasses
  • Can also be used for other laboratory items, such as filter discs
  • Easy to clean
Specifications
  • Type:
    Straight, standard quality
  • Length:
    115 mm
  • Seq:
    01