Double spatulas, micro, with one spoon end
185 mm, 5 mm
Catalog # RSGA395.185
Supplier: RSG
Jump to:
Product Details & Documents
Micro double spatulas with one spoon shaped end and one spatula end. Made of stainless steel.
Specifications
- Length:185 mm
- Blade width:5 mm



