Orders LinkContactUsLinkComponent
Laboratory knives
Laboratory knives
Broad blade, 120 mm
Catalog # 233-0148
Supplier:   RSG
Laboratory knives
Catalog # 233-0148
Supplier:  RSG
Laboratory knives
Laboratory knives
Broad blade, 120 mm
Price
182,00 €
Log in for availability
1 * 1 items
Product Details & Documents
These ceramic laboratory knives have extremely hard blades which resist chemicals, fractures and corrosion.
  • Anti-magnetic and lightweight
  • Dishwasher-proof
  • Long-life sharpness
Specifications
  • Type:
    Broad blade
  • Seq:
    03
  • Blade:
    120 mm