Laboratory knives
Thin blade, 90 mm
Catalog # 233-0146
Supplier: RSG
Jump to:
Product Details & Documents
These ceramic laboratory knives have extremely hard blades which resist chemicals, fractures and corrosion.
- Anti-magnetic and lightweight
- Dishwasher-proof
- Long-life sharpness
Specifications
- Type:Thin blade
- Seq:01
- Blade:90 mm