Order Entry
Export
Orders LinkContactUsLinkComponent
Sample loops for sample injectors, Rheodyne®

Sample loops for sample injectors, Rheodyne®

Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100, PEEK, 50 µl, 0,51 mm, 1,5 mm
Catalog # 554-2179
Supplier:   IDEX RHEODYNE
Sample loops for sample injectors, Rheodyne®
Catalog # 554-2179
Supplier:  IDEX RHEODYNE
Sample loops for sample injectors, Rheodyne®
Sample loops for sample injectors, Rheodyne®

Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100, PEEK, 50 µl, 0,51 mm, 1,5 mm

Price
114,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Capacity:
    50 µl
  • Description:
    Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100
  • Material:
    PEEK
  • Seq:
    26
  • For tube I-Ø:
    0,51 mm
  • For tube O-Ø:
    1,5 mm