Sample loops for sample injectors, Rheodyne®
Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100, PEEK, 5 µl, 0,18 mm, 1,5 mm
554-2176
- Capacity:5 µl
- Description:Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100
- Material:PEEK
- Seq:20
- For tube I-Ø:0,18 mm
- For tube O-Ø:1,5 mm



