Order Entry
Export
Orders LinkContactUsLinkComponent
Sample loops for sample injectors, Rheodyne®

Sample loops for sample injectors, Rheodyne®

Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100, Stainless steel, 2 ml, 1,00 mm, 1,5 mm
Catalog # 554-2162
Supplier:   IDEX RHEODYNE
Sample loops for sample injectors, Rheodyne®
Catalog # 554-2162
Supplier:  IDEX RHEODYNE
Sample loops for sample injectors, Rheodyne®
Sample loops for sample injectors, Rheodyne®

Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100, Stainless steel, 2 ml, 1,00 mm, 1,5 mm

Price
245,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Capacity:
    2 ml
  • Description:
    Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100
  • Material:
    Stainless steel
  • Seq:
    35
  • For tube I-Ø:
    1,00 mm
  • For tube O-Ø:
    1,5 mm