Order Entry
Export
Orders LinkContactUsLinkComponent
 

Sample loops for sample injectors, Rheodyne®

Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100, Stainless steel, 1 ml, 0,51 mm, 1,5 mm
  554-2159
Sample loops for sample injectors, Rheodyne®
  554-2159
Sample loops for sample injectors, Rheodyne®
Sample loops for sample injectors, Rheodyne®

Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100, Stainless steel, 1 ml, 0,51 mm, 1,5 mm

 
125,00 €
   
1 * 1 items
 
 
  • Capacity:
    1 ml
  • Description:
    Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100
  • Material:
    Stainless steel
  • Seq:
    32
  • For tube I-Ø:
    0,51 mm
  • For tube O-Ø:
    1,5 mm