Sample injectors, Rheodyne®
7725I 2/6 front loading manual injection valve with MBB™ * and position sensor, Stainless steel
Catalog # 554-2152
Supplier: IDEX RHEODYNE
Jump to:
Product Details & Documents
Manual sample injectors have two positions, 'Load' and 'Inject'. The 'Load' position allows sample loading into the sample loop or sample chamber. A syringe is commonly used to load sample into the sample injector. After loading, manual rotation of the sample injector's handle to the 'Inject' position allows the flushing of loaded sample onto a column.
* MBB™ (Make-Before-Break): Models incorporating Rheodyne's patented MBB architecture design provide uninterrupted flow when switching between 'Load' and 'Inject' positions.
Specifications
- Description:7725I 2/6 front loading manual injection valve with MBB™ * and position sensor
- Material:Stainless steel
- Seq:13
Accessories
Product Information
- Catalog #AvailabilityPrice
- 8125-999 RheBuild® kit for models 8125 and 81268125-999 RheBuild® kit for models 8125 and 8126554-2169Log in for availability1 * 1 items - 349,00 €
Product Information
- Catalog #AvailabilityPrice
- 9725-999 RheBuild® kit for models 9725 and 9725i9725-999 RheBuild® kit for models 9725 and 9725i554-2195Log in for availability1 * 1 items - 309,00 €