Order Entry
Export
Orders LinkContactUsLinkComponent
 

Sample loops for sample injectors, Rheodyne®

Sample loop, for model 7125, 7010, Stainless steel, 500 µl, 0,76 mm, 1,5 mm
Catalog # 554-2075
Supplier:   IDEX RHEODYNE
Sample loops for sample injectors, Rheodyne®
Catalog # 554-2075
Supplier:  IDEX RHEODYNE
Sample loops for sample injectors, Rheodyne®
Sample loops for sample injectors, Rheodyne®

Sample loop, for model 7125, 7010, Stainless steel, 500 µl, 0,76 mm, 1,5 mm

Price
101,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Capacity:
    500 µl
  • Description:
    Sample loop, for model 7125, 7010
  • Material:
    Stainless steel
  • Seq:
    51
  • For tube I-Ø:
    0,76 mm
  • For tube O-Ø:
    1,5 mm