Order Entry
Export
Orders LinkContactUsLinkComponent
Sample loops for sample injectors, Rheodyne®

Sample loops for sample injectors, Rheodyne®

Sample loop, for model 7125, 7010, Stainless steel, 100 µl, 0,51 mm, 1,5 mm
  554-2073
Sample loops for sample injectors, Rheodyne®
  554-2073
Sample loops for sample injectors, Rheodyne®
Sample loops for sample injectors, Rheodyne®

Sample loop, for model 7125, 7010, Stainless steel, 100 µl, 0,51 mm, 1,5 mm

 
123,00 €
   
1 * 1 items
 
 
  • Capacity:
    100 µl
  • Description:
    Sample loop, for model 7125, 7010
  • Material:
    Stainless steel
  • Seq:
    49
  • For tube I-Ø:
    0,51 mm
  • For tube O-Ø:
    1,5 mm