Orders LinkContactUsLinkComponent
Pierce™ ABC Staining Kits
Pierce™ ABC Staining Kits
ABC peroxidase standard staining kit, sufficient for at least 10 ml of ABC staining solution, 1 KIT
  PIER32020
Pierce™ ABC Staining Kits
  PIER32020
 :  
Pierce™ ABC Staining Kits
Pierce™ ABC Staining Kits
ABC peroxidase standard staining kit, sufficient for at least 10 ml of ABC staining solution, 1 KIT
 :  
496,00 €
1 * 1 KIT
ABC Staining Kits include reagents for the avidin-biotin complex (ABC) technique, a highly sensitive method for immunohistochemical detection with biotinylated secondary antibodies.
  • Very low background staining
  • Rapid avidin-biotin interaction
Standard Kits containing only the two essential reagents (avidin and biotinylated HRP) can be used with nearly any species of biotinylated secondary antibody. The Ultra-sensitive ABC Peroxidase Staining Kits enable more dilute primary antibodies to be used while producing equal staining intensity. Ultra-sensitive kits are offered as a standard kit and as complete kits, which contain biotinylated secondary antibodies and appropriate blocking serum for use with mouse or rabbit antibodies.
Documents
  • Pk:
    1 KIT
  • Description:
    ABC peroxidase standard staining kit, sufficient for at least 10 ml of ABC staining solution