Order Entry
Export
Orders LinkContactUsLinkComponent
 

Accessories for chromatography systems

Pre-flanged tubing, 1 m, Int.Ø 1,9 mm, Ext.Ø 2,7 mm, 2
Catalog # 18-8210-01
Supplier:   Cytiva
Accessories for chromatography systems
Catalog # 18-8210-01
Supplier:  Cytiva
CAS Number:  
Accessories for chromatography systems
Accessories for chromatography systems

Pre-flanged tubing, 1 m, Int.Ø 1,9 mm, Ext.Ø 2,7 mm, 2

CAS Number:  
Price
156,00 €
Log in for availability
1 * 2 items
Specifications

Specifications

  • Pk:
    2
  • Description:
    Pre-flanged tubing, 1 m, Int.Ø 1,9 mm, Ext.Ø 2,7 mm

Specifications