Order Entry
Export
Orders LinkContactUsLinkComponent
Accessories for chromatography systems

Accessories for chromatography systems

Unflanged tubing, 4 m, PTFE, Int.Ø 1,9 mm, Ext.Ø 2,7 mm, 7
Catalog # 18-8207-02
Supplier:   Cytiva
Accessories for chromatography systems
Catalog # 18-8207-02
Supplier:  Cytiva
CAS Number:  
Accessories for chromatography systems
Accessories for chromatography systems

Unflanged tubing, 4 m, PTFE, Int.Ø 1,9 mm, Ext.Ø 2,7 mm, 7

CAS Number:  
Price
492,00 €
Log in for availability
1 * 7 items
Specifications

Specifications

  • Pk:
    7
  • Description:
    Unflanged tubing, 4 m, PTFE, Int.Ø 1,9 mm, Ext.Ø 2,7 mm

Specifications