Orders LinkContactUsLinkComponent
Power Cables for Vacuum Controllers
Power Cables for Vacuum Controllers
Power cable KG, CH-plug
Catalog # VACU20676021
Supplier:   VACUUBRAND
Power Cables for Vacuum Controllers
Catalog # VACU20676021
Supplier:  VACUUBRAND
Power Cables for Vacuum Controllers
Power cable KG, CH-plug
Price
18,20 €
Log in for availability
1 * 1 items
Product Details & Documents
Power cables for Peltronic® exhaust vapour condenser and vacuum controllers VNC 2.
Specifications
  • Description:
    Power cable KG, CH-plug
  • Seq:
    07