Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Protein Purification
Protein Purification Equipment
Accessories for chromatography systems
Print
Accessories for chromatography systems
Union Luer lock female, M6 male, PP, 2
Catalog # 18-1027-62
Supplier:
Cytiva
Accessories for chromatography systems
Catalog # 18-1027-62
Supplier:
Cytiva
CAS Number:
10+
Return to Top
Accessories for chromatography systems
Union Luer lock female, M6 male, PP, 2
Specifications
Jump to:
Specifications
CAS Number:
Price
74,00 €
Log in
for availability
1 * 2 items
-
+
Other Product Family Options Available
Return Policy
Product Details & Documents
18100368_DATA FILE
18100368_DATA FILE 1
18102712_DATA FILE
18102712_DATA FILE 1
18321603_DATA FILE
18102712_HAND BOOK
18321603-HAND BOOK
18321603_INSTRUCTIONS
Cytiva CyDye Licensing Statements
Cytiva General Licensing Statements
Specifications
Specifications
Pk:
2
Description:
Union Luer lock female, M6 male, PP
Specifications