Orders LinkContactUsLinkComponent
Reducing pieces with small flanges KF
Reducing piece, DN 40/25, Stainless steel
Catalog # 185-2315
Supplier:   VACUUBRAND
Reducing pieces with small flanges KF
Catalog # 185-2315
Supplier:  VACUUBRAND
Reducing pieces with small flanges KF
Reducing piece, DN 40/25, Stainless steel
Price
95,50 €
Log in for availability
1 * 1 items
Product Details & Documents
Reducing pieces with small flanges KF made of aluminium or stainless steel.
Specifications
  • Description:
    Reducing piece, DN 40/25
  • Material:
    Stainless steel
  • Seq:
    10