Desalting columns, HiPrep™
HiPrep™ 26/10 Desalting, PP, 26×100 mm, 1×53 ml, ≤15 ml, <31 ml/min, <450 cm/h
Supplier: Cytiva
Jump to:
Product Details & Documents
HiPrep™ 26/10 desalting column is a convenient, easy to use column pre-packed with Sephadex™ G-25 Fine for desalting and buffer exchange with sample sizes of up to 15 ml per column.
- Replaces dialysis for desalting to saves time and reduces labour
- Compatible with a laboratory pump or chromatography system
- Two or four columns can be used in series for sample sizes of 30 to 60 ml
Documents
Certificates
Search by Lot Number17508701_DATA FILE 17508701_DATA FILE 1 17508701_DATA FILE 2 17508701_DATA FILE 3 17508702_DATA FILE 1.pdf
Datasheet
More
17508701_HAND BOOK 17508701_HAND BOOK 1 17508701_HAND BOOK 2 17508701_HAND BOOK 3 17508701_HAND BOOK 4
Manual
More
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Specifications
- Flow rate:<31 ml/min
- Capacity:1×53 ml
- Description:HiPrep™ 26/10 Desalting
- Seq:01
- Column hardware:PP
- Bed dimensions:26×100 mm
- Flow velocity:<450 cm/h
- Sample volume:≤15 ml
- Cat. No.:17-5087-01
- Max. pressure:1,5 bar
- Storage:4 to 30 °C, 20% Ethanol
- pH range:2 - 13



