Orders LinkContactUsLinkComponent
Mohr pinch cocks
Mohr pinch cocks
80 mm
Catalog # 229-0335
Supplier:   USBECK Laborgeräte
Mohr pinch cocks
Catalog # 229-0335
Mohr pinch cocks
Mohr pinch cocks
80 mm
Price
6,40 €
Log in for availability
1 * 1 items
Product Details & Documents
Nickel-plated brass. For regulating or stopping the flow of liquids in tube connections.
  • Very good thermal resistance
  • Easy handling
  • Easy cleaning
Specifications
  • Length:
    80 mm