Cork borer sets
Cork borer, 25,0 mm
Catalog # 217-0840
Supplier: USBECK Laborgeräte
Jump to:
Product Details & Documents
Brass, nickel plated, with handles and ejector.
- For manual boring of cork plugs
- Easy to clean
- Also available, cork drill sharpener compatible with all sets
Specifications
- Hole Ø:25,0 mm
- Description:Cork borer
- Seq:13
Accessories
Product Information
- Catalog #AvailabilityPrice
- Cork borer sharpener