Order Entry
Export
Orders LinkContactUsLinkComponent
Double spatulas for mortars

Double spatulas for mortars

150 mm, 20 mm
Catalog # 231-0448
Supplier:   USBECK Laborgeräte
Double spatulas for mortars
Catalog # 231-0448
Double spatulas for mortars
Double spatulas for mortars

150 mm, 20 mm

Price
4,10 €
Log in for availability
1 * 1 items
Product Details & Documents

Stainless steel.

  • Rigid
  • With two ends
Specifications
  • Length:
    150 mm
  • Blade width:
    20 mm