Orders LinkContactUsLinkComponent
Tweezers
Tweezers
Blunt, 160 mm
Catalog # 232-0090
Supplier:   USBECK Laborgeräte
Tweezers
Catalog # 232-0090
Tweezers
Tweezers
Blunt, 160 mm
Price
4,10 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel. A large range of quality forceps for various laboratory applications, production applications etc.
  • Practical
  • Anti-magnetic
  • Resistant to corrosives
Specifications
  • Type:
    Blunt
  • Length:
    160 mm