Burette clamps
Two burettes
Catalog # 241-7462
Supplier: USBECK Laborgeräte
Jump to:
Product Details & Documents
Die-cast aluminium. For simple and secure fixing of burettes.
- With bosshead for fixing to retort stand
- Span of clamp 0 to 20 mm
- With roll holders
Specifications
- For:Two burettes