Orders LinkContactUsLinkComponent
Burette clamps
One burette
Catalog # 241-7461
Supplier:   USBECK Laborgeräte
Burette clamps
Catalog # 241-7461
Burette clamps
Burette clamps
One burette
Price
26,60 €
Log in for availability
1 * 1 items
Product Details & Documents
Die-cast aluminium. For simple and secure fixing of burettes.
  • With bosshead for fixing to retort stand
  • Span of clamp 0 to 20 mm
  • With roll holders
Specifications
  • For:
    One burette