Clamps, three pronged
0 - 40 mm, Rod Ø 10 mm
Supplier: USBECK Laborgeräte
Jump to:
Product Details & Documents
Zinc die-cast, powder coated.
- The prongs of the clamps are covered with a plastic coating to protect the items clamped
- Without bosshead for fixing to stand
- With wing screws for rapid easy fixing
Documents
Certificates
Search by Lot NumberSpecifications
- For:Rod Ø 10 mm
- Spanning width:0 - 40 mm



